1,3-Benzodioxol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,2-Methylendioxybenzol
A14192.0B
A14192.18
A14192.30
A14192.0BEA
575
EUR
A14192.0B
A14192.18
A14192.30
1,3-Benzodioxol ≥99%
1,3-Benzodioxol
Purity: 99%
CAS No.: 274-09-9
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 55°(131°F)
Refractive Index: 1.5390
Odour: Aromatic
Melting Point: -18°
Boiling Point: 172-173°
Beilstein Reference: 115506
MDL No.: MFCD00005818
Formel:
C₇H₆O₂ Molecular Weight: 122.12 g/mol Siedepunkt: 172…173 °C Schmelzpunkt: –18 °C Dichte: 1.185 Flammpunkt: 55 °C (131 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005818 CAS-Nummer: 274-09-9 EINECS: 205-992-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...