D(+)-Biotin ≥98%
Lieferant: Thermo Fisher Scientific
Purity: 98+%
CAS No.: 58-85-5
Molecular Formula: C10H16N2O3S
Formula Weight: 244.32
Storage Temperature: 2°C to 8°C
Melting Point: 224-226°
Beilstein Reference: 86838
Merck Reference: 14,1231
Optical Rotation: +91° (c=1 in 0.1N NaOH)
MDL No.: MFCD00005541
Essential vitamin that is important for amino acid and energy metabolism
Formel:
C₁₀H₁₆N₂O₃S Schmelzpunkt: 224…226 °C Dichte: 1,41 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005541 CAS-Nummer: 58-85-5 EINECS: 200-399-3 Merck Index: 14,1231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...