Cyclopentanon ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A14222.0F A14222.AK A14222.AP
A14222.0FEA 166 EUR
A14222.0F A14222.AK A14222.AP
Cyclopentanon ≥99%
Cyclopentanone

Purity: 99% CAS No.: 120-92-3 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 30°(86°F) Refractive Index: 1.4370 Odour: Petrol-like Melting Point: -51° Boiling Point: 130-131° Beilstein Reference: 605573 Merck Reference: 14,2743 UN No.: 2245 MDL No.: MFCD00001409

Formel: C₅H₈O
Siedepunkt: 130…131 °C
Schmelzpunkt: –51 °C
Dichte: 0,949
Flammpunkt: 30 °C (86 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001409
CAS-Nummer: 120-92-3
EINECS: 204-435-9
UN: 2245
ADR: 3,III
Merck Index: 14,2743

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR