N-(2-Hydroxyethyl)phthalimid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(2-Hydroxyethyl)phthalsäureimid, N-(2-Hydroxyethyl)-1,2-benzoldicarbonsäureimid

A14259.22 A14259.36
A14259.22EA 47.5 EUR
A14259.22 A14259.36
N-(2-Hydroxyethyl)phthalimid ≥98%
N-(2-Hydroxyethyl)phthalimid

Purity: 98+% CAS No.: 3891-07-4 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Melting Point: 126-129° Beilstein Reference: 147214

Formel: C₁₀H₉NO₃
Schmelzpunkt: 126…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005903
CAS-Nummer: 3891-07-4
EINECS: 223-434-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR