cis-2-Penten-1-ol ≥97% remainder mainly trans-isomer
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(Z)-2-Penten-1-ol
A14262.06
A14262.14
A14262.22
A14262.06EA
52.5
EUR
A14262.06
A14262.14
A14262.22
cis-2-Penten-1-ol ≥97% remainder mainly trans-isomer
cis-2-Penten-1-ol
Purity: 97%
CAS No.: 1576-95-0
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 48°(118°F)
Refractive Index: 1.4360
Boiling Point: 138°
Beilstein Reference: 1719473
UN No.: 1987
MDL No.: MFCD00063208
Formel:
C₅H₁₀O Siedepunkt: 138 °C Dichte: 0,853 Flammpunkt: 48 °C (118 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063208 CAS-Nummer: 1576-95-0 EINECS: 216-415-7 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...