Magnesiumhydrogenorthophosphat Trihydrat ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Magnesiumphosphat dibasisch Trihydrat, Magnesiumorthophosphat dibasisch Trihydrat, Magnesiumhydrogenorthophosphat Trihydrat

A14279.0B
A14279.0BEA 60.6 EUR
A14279.0B
Magnesiumhydrogenorthophosphat Trihydrat ≥99%
Magnesiumhydrogenorthophosphat Trihydrat

Purity: 99% CAS No.: 7782-75-4 Molecular Formula: MgHPO4.3H2O Formula Weight: 174.34 (120.28anhy) Odour: Odourless Merck Reference: 14,5681 MDL No.: MFCD00150390

Formel: HMgO₄P·3H₂O
Dichte: 2,13
Lagertemperatur: Raumtemperatur
MDL: MFCD00150390
CAS-Nummer: 7782-75-4
EINECS: 231-823-5
Merck Index: 14,5681

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR