1,1-Cyclobutandicarbonsäure ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A14288.06 A14288.14 A14288.22
A14288.06EA 52.4 EUR
A14288.06 A14288.14 A14288.22
1,1-Cyclobutandicarbonsäure ≥99%
1,1-Cyclobutandicarbonsäure

Purity: 99% CAS No.: 5445-51-2 Molecular Formula: C6H8O4 Formula Weight: 144.13 Melting Point: ca 158° dec. Beilstein Reference: 2046031 UN No.: 3261 MDL No.: MFCD00001325

Formel: C₆H₈O₄
Siedepunkt: 663 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001325
CAS-Nummer: 5445-51-2
EINECS: 226-651-2
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR