Cyclohexyltriphenylphosphoniumbromid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14298.06
A14298.14
A14298.22
A14298.06EA
30.9
EUR
A14298.06
A14298.14
A14298.22
Cyclohexyltriphenylphosphoniumbromid ≥98%
Cyclohexyltriphenylphosphonium bromide
Purity: 98+%
CAS No.: 7333-51-9
Molecular Formula: C24H26BrP
Formula Weight: 425.36
Melting Point: 265-272°
Beilstein Reference: 3581012
Formel:
C₂₄H₂₆BrP Schmelzpunkt: 265…272 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031667 CAS-Nummer: 7333-51-9 EINECS: 230-834-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...