Cyclohexyltriphenylphosphoniumbromid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14298.06 A14298.14 A14298.22
A14298.06EA 30.9 EUR
A14298.06 A14298.14 A14298.22
Cyclohexyltriphenylphosphoniumbromid ≥98%
Cyclohexyltriphenylphosphonium bromide

Purity: 98+% CAS No.: 7333-51-9 Molecular Formula: C24H26BrP Formula Weight: 425.36 Melting Point: 265-272° Beilstein Reference: 3581012

Formel: C₂₄H₂₆BrP
Schmelzpunkt: 265…272 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031667
CAS-Nummer: 7333-51-9
EINECS: 230-834-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR