4-Brom-m-toluylsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Brom-3-methylbenzoesäure

A14302.06 A14302.14
A14302.06EA 70.6 EUR
A14302.06 A14302.14
4-Brom-m-toluylsäure ≥98%
4-Brom-m-toluylsäure

Purity: 98% CAS No.: 7697-28-1 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 215-218° Beilstein Reference: 2356649

Formel: C₈H₇BrO₂
Schmelzpunkt: 215…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039526
CAS-Nummer: 7697-28-1
EINECS: 231-713-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR