Methyl-3-(trifluormethyl)benzoat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 3-(trifluoromethyl)benzoate

A14316.06 A14316.14
A14316.06EA 31.4 EUR
A14316.06 A14316.14
Methyl-3-(trifluormethyl)benzoat ≥97%
Methyl-3-(trifluormethyl)benzoat

Purity: 97% CAS No.: 2557-13-3 Molecular Formula: C9H7F3O2 Formula Weight: 204.15 Flash Point: 84°(183°F) Refractive Index: 1.4530 Boiling Point: 80-82°/15mm Beilstein Reference: 1963428 MDL No.: MFCD00043543

Formel: C₉H₇F₃O₂
Molecular Weight: 204.15 g/mol
Siedepunkt: 80…82 °C (15 mmHg)
Dichte: 1.295
Flammpunkt: 84 °C (183 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00043543
CAS-Nummer: 2557-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR