N-Iodsuccinimid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14320.06
A14320.22
A14320.14
AB109218.0100
A14320.06EA
48
EUR
A14320.06
A14320.22
A14320.14
ABCRAB109218.0100
N-Iodsuccinimid ≥97%
N-Iodsuccinimid
Purity: 97%
CAS No.: 516-12-1
Molecular Formula: C4H4INO2
Formula Weight: 224.99
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Density: 2.245
Sensitivity: Moisture and Light Sensitive
Appearance: White to pale brown
Melting Point: ca 196° dec.
Beilstein Reference: 113917
Merck Reference: 14,5045
MDL No.: MFCD00005512
Formel:
C₄H₄INO₂ Siedepunkt: 356 °C (1013 hPa) Schmelzpunkt: ∼ 196 °C (Zersetzung) Dichte: 2,245 Lagertemperatur: Kühlschrank |
MDL:
MFCD00005512 CAS-Nummer: 516-12-1 EINECS: 208-221-6 Merck Index: 14,5045 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...