1-(Cyanmethyl)pyridiniumchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A14327.09 A14327.18
A14327.09EA 38.1 EUR
A14327.09 A14327.18
1-(Cyanmethyl)pyridiniumchlorid ≥97%
1-(Cyanomethyl)pyridinium chloride

Purity: 97% CAS No.: 17281-59-3 Molecular Formula: C7H7ClN2 Formula Weight: 154.60 Melting Point: 173-175° Beilstein Reference: 3728392 UN No.: 3439 MDL No.: MFCD00031933

Formel: C₇H₇ClN₂
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031933
CAS-Nummer: 17281-59-3
EINECS: 241-312-9
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR