5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB) ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14331.03
A14331.06
A14331.14
A14331.03EA
26.5
EUR
A14331.03
A14331.06
A14331.14
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB) ≥99%
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Purity: 99%
CAS No.: 69-78-3
Molecular Formula: C14H8N2O8S2
Formula Weight: 396.36
Melting Point: ca 240° dec.
Beilstein Reference: 1896821
Formel:
C₁₄H₈N₂O₈S₂ Schmelzpunkt: ∼ 240 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007140 CAS-Nummer: 69-78-3 EINECS: 200-714-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...