2,3-Butandionmonoxim ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Diacetylmonoxim
A14339.0E
A14339.22
A14339.36
A14339.0EEA
1023
EUR
A14339.0E
A14339.22
A14339.36
2,3-Butandionmonoxim ≥99%
2,3-Butandionmonoxim
Purity: 99%
CAS No.: 57-71-6
Molecular Formula: C4H7NO2
Formula Weight: 101.11
Storage Temperature: 2°C to 8°C
Melting Point: 74-77°
Boiling Point: 185-186°
Beilstein Reference: 605582
MDL No.: MFCD00002116
Formel:
C₄H₇NO₂ Siedepunkt: 185…186 °C Schmelzpunkt: 74…77 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00002116 CAS-Nummer: 57-71-6 EINECS: 200-348-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...