3-Fluor-4-nitrophenol ≥99%

Lieferant: Thermo Scientific
Gefahr

A14350.06
A14350.06EA 106 EUR
A14350.06
3-Fluor-4-nitrophenol ≥99%
3-Fluor-4-nitrophenol

Purity: 99% CAS No.: 394-41-2 Molecular Formula: C6H4FNO3 Formula Weight: 157.10 Melting Point: 91-93° Beilstein Reference: 2048033

Formel: C₆H₄FNO₃
Molecular Weight: 157,1 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041251
CAS-Nummer: 394-41-2
EINECS: 206-895-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR