Acetanilid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14361.0E A14361.22 A14361.36
A14361.0EEA 211 EUR
A14361.0E A14361.22 A14361.36
Acetanilid ≥98%
Acetanilid

Purity: 98% CAS No.: 103-84-4 Molecular Formula: C8H9NO Formula Weight: 135.17 Flash Point: 161°(321°F) Physical Form: Crystals Density: 1.121 Melting Point: 113-116° Boiling Point: 304-305° Beilstein Reference: 606468 Merck Reference: 14,50 MDL No.: MFCD00008674

Formel: C₈H₉NO
Siedepunkt: 304…305 °C
Schmelzpunkt: 113…116 °C
Dichte: 1,121
Flammpunkt: 161 °C (321 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008674
CAS-Nummer: 103-84-4
EINECS: 203-150-7
Merck Index: 14,50

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR