4-(Hydroxymethyl)benzylalkohol ≥99%

Lieferant: Thermo Fisher Scientific

A14405.06 A14405.14 A14405.22 AB113850.0005
A14405.06EA 51.4 EUR
A14405.06 A14405.14 A14405.22 ABCRAB113850.0005
4-(Hydroxymethyl)benzylalkohol ≥99%
4-(Hydroxymethyl)benzylalkohol

Purity: 99% CAS No.: 589-29-7 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 117-121° Boiling Point: 138-143°/1mm MDL No.: MFCD00004665

Formel: C₈H₁₀O₂
Siedepunkt: 138…143 °C (1 mmHg)
Schmelzpunkt: 117…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004665
CAS-Nummer: 589-29-7
EINECS: 209-641-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR