2-Nitro-4-(trifluormethyl)anilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A14413.09 A14413.18 A14413.30
A14413.09EA 41.9 EUR
A14413.09 A14413.18 A14413.30
2-Nitro-4-(trifluormethyl)anilin ≥98%
2-Nitro-4-(trifluormethyl)anilin

Purity: 98% CAS No.: 400-98-6 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 105-109° Beilstein Reference: 650808 UN No.: 2811 MDL No.: MFCD00007155

Formel: C₇H₅F₃N₂O₂
Schmelzpunkt: 105…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007155
CAS-Nummer: 400-98-6
EINECS: 206-926-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR