1-Butyl-4-iodbenzol ≥98%

Lieferant: Thermo Scientific

Synonyme: 1-(4'-Iodphenyl)butan, 1-(4-Iodphenyl)butan

A14421.14
A14421.14EA 113 EUR
A14421.14
1-Butyl-4-iodbenzol ≥98%
1-Butyl-4-iodbenzol

Purity: 98% CAS No.: 20651-67-6 Molecular Formula: C10H13I Formula Weight: 260.12 Refractive Index: 1.5690 Boiling Point: 80-82°/0.5mm MDL No.: MFCD00051927

Formel: C₁₀H₁₃I
Molecular Weight: 260,12 g/mol
Siedepunkt: 80…82 °C (0,5 mmHg)
Dichte: 1,462
Lagertemperatur: Raumtemperatur
MDL: MFCD00051927
CAS-Nummer: 20651-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR