2,4-Dibrom-6-fluoranilin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
A14435.14
A14435.14EA
65
EUR
A14435.14
2,4-Dibrom-6-fluoranilin ≥97%
2,4-Dibrom-6-fluoranilin
Purity: 97%
CAS No.: 141474-37-5
Molecular Formula: C6H4Br2FN
Formula Weight: 268.92
Melting Point: 62-64°
Beilstein Reference: 6289292
UN No.: 2811
MDL No.: MFCD00042230
Formel:
C₆H₄Br₂FN Molecular Weight: 268,91 g/mol Siedepunkt: 277 °C (1013 hPa) Schmelzpunkt: 62…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042230 CAS-Nummer: 141474-37-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...