Bromwasserstoff-Lösung 33% (w/w) 45% (w/v) in Essigsäure

Lieferant: Thermo Fisher Scientific
Gefahr

A14475.0F A14475.AE A14475.AP
A14475.0FEA 414 EUR
A14475.0F A14475.AE A14475.AP
Bromwasserstoff-Lösung 33% (w/w) 45% (w/v) in Essigsäure
Bromwasserstoff

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds


It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

Formel: HBr
Dichte: 1,40
Flammpunkt: 65 °C (149 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011323
CAS-Nummer: 37348-16-6
EINECS: 233-113-0
UN: 3265
ADR: 8,II
Merck Index: 14,4778

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR