2,6-Xylidin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-m-xylol, 2,6-Dimethylanilin

A14512.0B A14512.22 A14512.36
A14512.0BEA 110 EUR
A14512.0B A14512.22 A14512.36
2,6-Xylidin ≥99%
2,6-Xylidin

Purity: 99% CAS No.: 87-62-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 91°(195°F) Refractive Index: 1.5600 Odour: Characteristic Melting Point: 11° Boiling Point: 216° Beilstein Reference: 636332 Merck Reference: 14,10084 UN No.: 1711 MDL No.: MFCD00007747

Formel: C₈H₁₁N
Molecular Weight: 121.18 g/mol
Siedepunkt: 216 °C
Schmelzpunkt: 11 °C
Dichte: 0.981
Flammpunkt: 91 °C (195 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007747
CAS-Nummer: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1 II
Merck Index: 12,10217

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR