α,α,α-Trifluor-p-toluonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Cyanbenzotrifluorid, 4-(Trifluormethyl)benzonitril, p-(Trifluormethyl)benzonitril, 4-(Trifluormethyl)benzoesäurenitril
A14514.09
A14514.18
A14514.30
A14514.09EA
44
EUR
A14514.09
A14514.18
A14514.30
α,α,α-Trifluor-p-toluonitril ≥98%
α,α,α-Trifluor-p-toluonitril
Purity: 98%
CAS No.: 455-18-5
Molecular Formula: C8H4F3N
Formula Weight: 171.12
Flash Point: 71°(159°F)
Refractive Index: 1.4583
Melting Point: 36-40°
Boiling Point: 183-185°
Beilstein Reference: 2046478
UN No.: 3439
MDL No.: MFCD00001826
Formel:
C₈H₄F₃N Siedepunkt: 183…185 °C Schmelzpunkt: 36…40 °C Dichte: 1,278 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001826 CAS-Nummer: 455-18-5 EINECS: 207-239-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...