N-(4-Brombutyl)phthalimid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(4-Brombutyl)-1,2-benzoldicarbonsäureimid, N-(4-Brombutyl)phthalsäureimid
A14517.06
A14517.14
A14517.22
A14517.06EA
54.9
EUR
A14517.06
A14517.14
A14517.22
N-(4-Brombutyl)phthalimid ≥98%
N-(4-Brombutyl)phthalimid
Purity: 98%
CAS No.: 5394-18-3
Molecular Formula: C12H12BrNO2
Formula Weight: 282.14
Odour: Odourless
Melting Point: 77-81°
Boiling Point: 165-170°/1mm
Beilstein Reference: 164119
MDL No.: MFCD00005905
Formel:
C₁₂H₁₂BrNO₂ Siedepunkt: 165…170 °C (1 mmHg) Schmelzpunkt: 77…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005905 CAS-Nummer: 5394-18-3 EINECS: 226-401-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...