N-(4-Brombutyl)phthalimid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(4-Brombutyl)-1,2-benzoldicarbonsäureimid, N-(4-Brombutyl)phthalsäureimid

A14517.06 A14517.14 A14517.22
A14517.06EA 54.9 EUR
A14517.06 A14517.14 A14517.22
N-(4-Brombutyl)phthalimid ≥98%
N-(4-Brombutyl)phthalimid

Purity: 98% CAS No.: 5394-18-3 Molecular Formula: C12H12BrNO2 Formula Weight: 282.14 Odour: Odourless Melting Point: 77-81° Boiling Point: 165-170°/1mm Beilstein Reference: 164119 MDL No.: MFCD00005905

Formel: C₁₂H₁₂BrNO₂
Siedepunkt: 165…170 °C (1 mmHg)
Schmelzpunkt: 77…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005905
CAS-Nummer: 5394-18-3
EINECS: 226-401-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR