4-Chlorphenylhydraziniumchlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14518.06
A14518.14
A14518.22
A14518.06EA
28.7
EUR
A14518.06
A14518.14
A14518.22
4-Chlorphenylhydraziniumchlorid ≥97%
4-Chlorphenylhydraziniumchlorid
Purity: 97%
CAS No.: 1073-70-7
Molecular Formula: C6H7ClN2.HCl
Formula Weight: 179.05
Odour: Odourless
Melting Point: ca 216° dec.
Beilstein Reference: 3596254
MDL No.: MFCD00012943
Formel:
C₆H₈Cl₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012943 CAS-Nummer: 1073-70-7 EINECS: 214-030-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...