2-Hydroxypyridin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A14522.14 A14522.22 A14522.36
A14522.14EA 31.7 EUR
A14522.14 A14522.22 A14522.36
2-Hydroxypyridin ≥98%

Purity: 98% CAS No.: 142-08-5 Molecular Formula: C5H5NO Formula Weight: 95.10 Storage Temperature: 2°C to 8°C Flash Point: 210°(410°F) Odour: Recognizable Melting Point: 102-111° Boiling Point: 280-281° Beilstein Reference: 105786 UN No.: 2811 MDL No.: MFCD00006268

Formel: C₅H₅NO
Siedepunkt: 280…281 °C
Schmelzpunkt: 102…111 °C
Dichte: 1,39
Flammpunkt: 210 °C (410 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00006268
CAS-Nummer: 142-08-5
EINECS: 205-520-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR