3-(4-Hydroxyphenyl)propionsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(4-Hydroxyphenyl)propansäure
A14567.09
A14567.18
A14567.30
A14567.09EA
33.9
EUR
A14567.09
A14567.18
A14567.30
3-(4-Hydroxyphenyl)propionsäure ≥99%
3-(4-Hydroxyphenyl)propionsäure
Purity: 99%
CAS No.: 501-97-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 126-131°
Beilstein Reference: 2209841
Formel:
C₉H₁₀O₃ Siedepunkt: 208…210 °C (14 mmHg) Schmelzpunkt: 126…131 °C Dichte: 1,336 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002778 CAS-Nummer: 501-97-3 EINECS: 207-931-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...