2'-Hydroxy-4',6'-dimethoxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-on

A14585.03 A14585.06
A14585.03EA 48.3 EUR
A14585.03 A14585.06
2'-Hydroxy-4',6'-dimethoxyacetophenon ≥98%
2'-Hydroxy-4',6'-dimethoxyacetophenon

Purity: 98% CAS No.: 90-24-4 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 80-83° Boiling Point: 185°/20mm Merck Reference: 14,10067 MDL No.: MFCD00017243

Formel: C₁₀H₁₂O₄
Molecular Weight: 196.2
Siedepunkt: 185 °C (20 mmHg)
Schmelzpunkt: 80…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017243
CAS-Nummer: 90-24-4
EINECS: 201-978-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR