Natriumtrifluoracetat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Trifluoressigsäure Natriumsalz

A14613.14 A14613.22 A14613.36
A14613.14EA 38.1 EUR
A14613.14 A14613.22 A14613.36
Natriumtrifluoracetat ≥98%
Natriumtrifluoracetat

Purity: 98% CAS No.: 2923-18-4 Molecular Formula: CF3CO2Na Formula Weight: 136.00 Melting Point: ca 207° dec. Beilstein Reference: 3597949

Formel: C₂F₃NaO₂
Molecular Weight: 136.01 g/mol
Schmelzpunkt: ∼ 207 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013217
CAS-Nummer: 2923-18-4
EINECS: 220-879-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR