N-Phenylmaleimid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A14616.14 A14616.22
A14616.14EA 54.4 EUR
A14616.14 A14616.22
N-Phenylmaleimid ≥98%
N-Phenylmaleimid

Purity: 98+% CAS No.: 941-69-5 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 87-91° Boiling Point: 162-163°/12mm Merck Reference: 14,7299 UN No.: 2811 MDL No.: MFCD00005502

Formel: C₁₀H₇NO₂
Siedepunkt: 162…163 °C (12 mmHg)
Schmelzpunkt: 87…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005502
CAS-Nummer: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,7299

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR