2',6'-Dihydroxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2,6-Dihydroxyphenyl)ethan-1-on

A14627.06 A14627.14 A14627.22
A14627.06EA 31.7 EUR
A14627.06 A14627.14 A14627.22
2',6'-Dihydroxyacetophenon ≥98%
2',6'-Dihydroxyacetophenon

Purity: 98+% CAS No.: 699-83-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 156-159° Beilstein Reference: 1366061

Formel: C₈H₈O₃
Molecular Weight: 152.15 g/mol
Siedepunkt: 296
Schmelzpunkt: 156…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002270
CAS-Nummer: 699-83-2
EINECS: 211-833-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR