3-Octen-2-on 97%
Lieferant: Thermo Fisher Scientific
Purity: 97%
CAS No.: 1669-44-9
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 54°(129°F)
Refractive Index: 1.4480
Odour: Pleasant
Boiling Point: 60°/16mm
Beilstein Reference: 1742230
UN No.: UN1224
MDL No.: MFCD00015565
Formel:
C₈H₁₄O |
MDL:
MFCD00015565 CAS-Nummer: 1669-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...