Methyl-3,4-diaminobenzoat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,4-Diaminobenzoesäuremethylester

A14656.06 A14656.14
A14656.06EA 64 EUR
A14656.06 A14656.14
Methyl-3,4-diaminobenzoat 98%
Methyl-3,4-diaminobenzoat

Purity: 98% CAS No.: 36692-49-6 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 105-108° Beilstein Reference: 2091675

Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.18 g/mol
Siedepunkt: 319
Schmelzpunkt: 105…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017098
CAS-Nummer: 36692-49-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR