1-Brom-4-octylbenzol ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
p-Brom-octylbenzol
A14676.03
A14676.06
A14676.03EA
39.1
EUR
A14676.03
A14676.06
1-Brom-4-octylbenzol ≥97%
1-Brom-4-octylbenzol
Purity: 97%
CAS No.: 51554-93-9
Molecular Formula: C14H21Br
Formula Weight: 269.23
MDL No.: MFCD00038348
Formel:
C₁₄H₂₁Br Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038348 CAS-Nummer: 51554-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...