1-Brom-4-octylbenzol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: p-Brom-octylbenzol

A14676.03 A14676.06
A14676.03EA 39.1 EUR
A14676.03 A14676.06
1-Brom-4-octylbenzol ≥97%
1-Brom-4-octylbenzol

Purity: 97% CAS No.: 51554-93-9 Molecular Formula: C14H21Br Formula Weight: 269.23 MDL No.: MFCD00038348

Formel: C₁₄H₂₁Br
Lagertemperatur: Raumtemperatur
MDL: MFCD00038348
CAS-Nummer: 51554-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR