6-Benzylaminopurin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Benzyl(purin-6-yl)amin, 6-Benzyladenin, N6-Benzyladenin
A14678.03
A14678.06
A14678.14
A14678.03EA
28.7
EUR
A14678.03
A14678.06
A14678.14
6-Benzylaminopurin ≥99%
6-Benzylaminopurin
Purity: 99%
CAS No.: 1214-39-7
Molecular Formula: C12H11N5
Formula Weight: 225.26
Melting Point: 229-233°
Beilstein Reference: 19406
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables
Formel:
C₁₂H₁₁N₅ Schmelzpunkt: 229…233 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005572 CAS-Nummer: 1214-39-7 EINECS: 214-927-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...