1,3-Dibrompropan ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Trimethylenbromid
A14687.30
A14687.0B
A14687.0I
AB109197.1000
A14687.30EA
50.8
EUR
A14687.30
A14687.0B
A14687.0I
ABCRAB109197.1000
1,3-Dibrompropan ≥98%
1,3-Dibrompropan
Purity: 98%
CAS No.: 109-64-8
Molecular Formula: C3H6Br2
Formula Weight: 201.90
Flash Point: 54°(130°F)
Physical Form: Liquid
Density: 1.99
Refractive Index: 1.5230
Appearance: Colourless
Melting Point: -34°
Boiling Point: 167°
Beilstein Reference: 635662
Merck Reference: 14,9713
UN No.: 1993
MDL No.: MFCD00000255
Formel:
BrH₂CCH₂CH₂Br Molecular Weight: 201.89 g/mol Siedepunkt: 167 °C Schmelzpunkt: –34 °C Dichte: 1.99 Flammpunkt: 54 °C (130 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000255 CAS-Nummer: 109-64-8 EINECS: 203-690-3 UN: 1993 ADR: 3 III Merck Index: 12,09844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...