1,1'-Carbonyldiimidazol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 530-62-1
Molecular Formula: C7H6N4O
Formula Weight: 162.15
Physical Form: Cryst powder
Sensitivity: Moisture Sensitive
Appearance: White to cream
pH: 7.7
Melting Point: 116-120°
Beilstein Reference: 6826
Merck Reference: 14,1819
UN No.: 3263
MDL No.: MFCD00005286
Peptide coupling reagent
Formel:
C₇H₆N₄O Schmelzpunkt: 116…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005286 CAS-Nummer: 530-62-1 EINECS: 208-488-9 UN: 3263 ADR: 8,II Merck Index: 14,1819 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...