trans-Zimtaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98+%
CAS No.: 14371-10-9
Molecular Formula: C9H8O
Formula Weight: 132.16
Flash Point: 131°(268°F)
Physical Form: Liquid
Density: 1.050
Refractive Index: 1.6220
Sensitivity: Air Sensitive
Appearance: Yellow
Odour: Characteristic
Melting Point: -8°
Boiling Point: 250-252°
Beilstein Reference: 1071571
Merck Reference: 14,2297
MDL No.: MFCD00007000
Formel:
C₉H₈O Siedepunkt: 250…252 °C Schmelzpunkt: –8 °C Dichte: 1,050 Flammpunkt: 131 °C (268 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007000 CAS-Nummer: 14371-10-9 Merck Index: 14,2297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...