Tricin ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(Tris(hydroxymethyl)methyl)glycin
A14695.14
A14695.22
A14695.36
A14695.14EA
26.3
EUR
A14695.14
A14695.22
A14695.36
Tricin ≥98%
Tricin
Purity: 98+%
CAS No.: 5704-04-1
Molecular Formula: (HOCH2)3CNHCH2CO2H
Formula Weight: 179.17
Melting Point: ca 183° dec.
Beilstein Reference: 1937804
Merck Reference: 14,9651
MDL No.: MFCD00004277
Formel:
(HOCH₂)₃CNHCH₂CO₂H Schmelzpunkt: ∼ 183 °C (Zersetzung) Dichte: 1,523 g/cm³ (19.85 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004277 CAS-Nummer: 5704-04-1 EINECS: 227-193-6 Merck Index: 14,9651 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...