2-Brom-5-nitrothiazol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14701.06 A14701.14
A14701.06EA 72.2 EUR
A14701.06 A14701.14
2-Brom-5-nitrothiazol ≥98%
2-Bromo-5-nitrothiazole

Purity: 98% CAS No.: 3034-48-8 Molecular Formula: C3HBrN2O2S Formula Weight: 209.03 Melting Point: 85-90° Beilstein Reference: 4856

Formel: C₃HBrN₂O₂S
Molecular Weight: 209.02 g/mol
Schmelzpunkt: 85…90 °C
Dichte: 2.295 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005317
CAS-Nummer: 3034-48-8
EINECS: 221-226-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR