Di-tert-butylpyrocarbonat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97+%
CAS No.: 24424-99-5
Molecular Formula: C10H18O5
Formula Weight: 218.25
Storage Temperature: 2°C to 8°C
Flash Point: 37°(99°F)
Physical Form: Low-melting solid
Density: 0.950
Refractive Index: 1.4090
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odour: Mild
Melting Point: 21-24°
Boiling Point: 56-57°/0.5mm
Beilstein Reference: 1911173
UN No.: 2930
MDL No.: MFCD00008805
Reagent for the introduction of the Boc protecting group
Formel:
((H₃C)₃COC(O))₂O Siedepunkt: 56…57 °C (0,5 mmHg) Schmelzpunkt: 21…24 °C Dichte: 0,950 Flammpunkt: 37 °C (99 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00008805 CAS-Nummer: 24424-99-5 EINECS: 246-240-1 UN: 2930 ADR: 6.1,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...