Hexanophenon ≥98%

Lieferant: Thermo Fisher Scientific

A14712.06 A14712.14 A14712.22
A14712.06EA 27.2 EUR
A14712.06 A14712.14 A14712.22
Hexanophenon ≥98%
Hexanophenon

Purity: 98% CAS No.: 942-92-7 Molecular Formula: C12H16O Formula Weight: 176.26 Flash Point: 110°(230°F) Refractive Index: 1.5105 Melting Point: 25-27° Boiling Point: 265° Beilstein Reference: 1908667 MDL No.: MFCD00009512

Formel: C₁₂H₁₆O
Siedepunkt: 265 °C
Schmelzpunkt: 25…27 °C
Dichte: 0,950
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009512
CAS-Nummer: 942-92-7
EINECS: 213-394-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR