Diethylenglycol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,2'-Dihydroxidiethylether, 2,2'-Oxydiethanol, Diglycol, Digol
A14728.0C
A14728.0E
A14728.30
A14728.36
AB115885.0250
A14728.0CEA
209
EUR
A14728.0C
A14728.0E
A14728.30
A14728.36
ABCRAB115885.0250
Diethylenglycol ≥99%
Diethylenglycol
Purity: 99%
CAS No.: 111-46-6
Molecular Formula: C4H10O3
Formula Weight: 106.12
Flash Point: 143°(289°F)
Physical Form: Clear liquid
Density: 1.118
Refractive Index: 1.4470
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Nearly Odourless
Melting Point: -10°
Boiling Point: 245-246°
Beilstein Reference: 969209
Merck Reference: 14,3119
MDL No.: MFCD00002882
Formel:
C₄H₁₀O₃ Molecular Weight: 106.12 g/mol Siedepunkt: 245…246 °C Schmelzpunkt: –10 °C Dichte: 1.118 Flammpunkt: 143 °C (289 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002882 CAS-Nummer: 111-46-6 EINECS: 203-872-2 Merck Index: 13,03146 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...