Diethylenglycol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2'-Dihydroxidiethylether, 2,2'-Oxydiethanol, Diglycol, Digol

A14728.0C A14728.0E A14728.30 A14728.36 AB115885.0250
A14728.0CEA 209 EUR
A14728.0C A14728.0E A14728.30 A14728.36 ABCRAB115885.0250
Diethylenglycol ≥99%
Diethylenglycol

Purity: 99% CAS No.: 111-46-6 Molecular Formula: C4H10O3 Formula Weight: 106.12 Flash Point: 143°(289°F) Physical Form: Clear liquid Density: 1.118 Refractive Index: 1.4470 Sensitivity: Hygroscopic Appearance: Colourless Odour: Nearly Odourless Melting Point: -10° Boiling Point: 245-246° Beilstein Reference: 969209 Merck Reference: 14,3119 MDL No.: MFCD00002882

Formel: C₄H₁₀O₃
Molecular Weight: 106.12 g/mol
Siedepunkt: 245…246 °C
Schmelzpunkt: –10 °C
Dichte: 1.118
Flammpunkt: 143 °C (289 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002882
CAS-Nummer: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR