trans-3-(3-Pyridyl)acrylic acid 99%

Lieferant: Thermo Fisher Scientific
Achtung

A14747.06 A14747.14 A14747.30
A14747.06EA 36.9 EUR
A14747.06 A14747.14 A14747.30
trans-3-(3-Pyridyl)acrylic acid 99%
trans-3-(3-Pyridyl)acrylic acid

Purity: 99% CAS No.: 19337-97-4 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Melting Point: 232-235° Beilstein Reference: 113290 MDL No.: MFCD00006410

Formel: C₈H₇NO₂
Schmelzpunkt: 232…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006410
CAS-Nummer: 19337-97-4
EINECS: 214-424-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR