1-Formylpiperazin ≥90%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Piperazinecarboxaldehyde, 1-Piperazinecarbaldehyde
A14757.14
A14757.22
A14757.36
A14757.14EA
69.9
EUR
A14757.14
A14757.22
A14757.36
1-Formylpiperazin ≥90%
1-Formylpiperazin
Purity: 90+%
CAS No.: 7755-92-2
Molecular Formula: C5H10N2O
Formula Weight: 114.15
Storage Temperature: 2°C to 8°C
Flash Point: 102°(216°F)
Refractive Index: 1.5120
Boiling Point: 264-266°
Beilstein Reference: 110295
MDL No.: MFCD00005963
Formel:
C₅H₁₀N₂O Molecular Weight: 114.15 g/mol Siedepunkt: 264…266 °C Dichte: 1.111 Flammpunkt: 102 °C (216 °F) |
MDL:
MFCD00005963 CAS-Nummer: 7755-92-2 EINECS: 231-813-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...