Methylendiphosphonsäure ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Medronic acid

A14803.03 A14803.06 A14803.14
A14803.03EA 70.5 EUR
A14803.03 A14803.06 A14803.14
Methylendiphosphonsäure ≥99%
Methylenediphosphonic acid

Purity: 99+% CAS No.: 1984-15-2 Molecular Formula: CH6O6P2 Formula Weight: 176.00 Odour: Odourless Melting Point: 202-204° Beilstein Reference: 1708494 Merck Reference: 14,5792 UN No.: 3261 MDL No.: MFCD00002139

Formel: CH₆O₆P₂
Molecular Weight: 176 g/mol
Schmelzpunkt: 202…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002139
CAS-Nummer: 1984-15-2
EINECS: 217-851-0
UN: 3261
ADR: 8 III
Merck Index: 14,5792

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR